siRNA Details
| Gene Symbol: 1700067K01Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK138628.1 | - | 1 | 16 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY724382.1 | - | 1 | 13 | Chr. 8 - 84004235 - 84004775 [-] on Build GRCm38 | |

1700067K01Rik
Genomic Map