siRNA Details
| Gene Symbol: 1810012K08Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK007459.1 | - | 1 | 13 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY707528.1 | - | 1 | 13 | Chr. 2 - 26138546 - 26139543 [+] on Build GRCm38 | |

1810012K08Rik
Genomic Map