siRNA Details
| Gene Symbol: 9330178D15Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040553.1 | 5 | 1568 | |||
| GenBank mRNA | AK034327.1 | - | 5 | 1569 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

9330178D15Rik| Gene Symbol: 9330178D15Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040553.1 | 5 | 1568 | |||
| GenBank mRNA | AK034327.1 | - | 5 | 1569 | ||