siRNA Details
Gene Symbol: Gm14204 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040358.1 | 3 | 4269 | |||
GenBank mRNA | AK034893.1 | - | 1 | 1234 | ||
BC072662.1 | - | 3 | 4269 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gm14204 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040358.1 | 3 | 4269 | |||
GenBank mRNA | AK034893.1 | - | 1 | 1234 | ||
BC072662.1 | - | 3 | 4269 |