siRNA Details
| Gene Symbol: Gm14204 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040358.1 | 3 | 3355 | |||
| GenBank mRNA | AK034893.1 | - | 1 | 320 | ||
| BC072662.1 | - | 3 | 3355 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm14204| Gene Symbol: Gm14204 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040358.1 | 3 | 3355 | |||
| GenBank mRNA | AK034893.1 | - | 1 | 320 | ||
| BC072662.1 | - | 3 | 3355 | |||