siRNA Details
| Gene Symbol: 1700021N21Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045800.1 | 3 | 879 | |||
| GenBank mRNA | AK006224.1 | - | 3 | 880 | ||
| BB686792.1 | - | 1 | 396 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1700021N21Rik| Gene Symbol: 1700021N21Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045800.1 | 3 | 879 | |||
| GenBank mRNA | AK006224.1 | - | 3 | 880 | ||
| BB686792.1 | - | 1 | 396 | |||