siRNA Details
| Gene Symbol: Gm38811 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK031323.1 | - | 471 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB620696.1 | - | 2 | 471 | Chr. 6 - 51697227 - 51711081 [-] on Build GRCm38 | |

Gm38811
Genomic Map