siRNA Details
| Gene Symbol: 7530416G11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK078701.1 | - | 5 | 715 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

7530416G11Rik| Gene Symbol: 7530416G11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK078701.1 | - | 5 | 715 | ||