siRNA Details
| Gene Symbol: 9530027D23Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK020577.1 | - | 1 | 375 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB485726.2 | - | 1 | 306 | Chr. 13 - 107852002 - 107852672 [-] on Build GRCm38 | |

9530027D23Rik
Genomic Map