siRNA Details
| Gene Symbol: 9530025L08Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK020572.1 | - | 1 | 536 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY722053.1 | - | 1 | 536 | Chr. 19 - 30165114 - 30165769 [-] on Build GRCm38 | |

9530025L08Rik
Genomic Map