siRNA Details
| Gene Symbol: 1700039M10Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK006320.1 | - | 2 | 93 | ||
| AK006644.1 | - | 2 | 82 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1700039M10Rik| Gene Symbol: 1700039M10Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK006320.1 | - | 2 | 93 | ||
| AK006644.1 | - | 2 | 82 | |||