siRNA Details
| Gene Symbol: 4930535I16Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK050163.1 | - | 1 | 1198 | ||
| AK085801.1 | - | 1 | 1222 | |||
| BB667541.1 | - | 1 | 602 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | CF723499.1 | - | 1 | 309 | Chr. 4 - 123915770 - 123916385 [-] on Build GRCm38 | |

4930535I16Rik
Genomic Map