siRNA Details
| Gene Symbol: 4930535I16Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK050163.1 | - | 1 | 779 | ||
| AK085801.1 | - | 1 | 803 | |||
| BB667541.1 | - | 1 | 183 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4930535I16Rik| Gene Symbol: 4930535I16Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK050163.1 | - | 1 | 779 | ||
| AK085801.1 | - | 1 | 803 | |||
| BB667541.1 | - | 1 | 183 | |||