siRNA Details
| Gene Symbol: Gm36594 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK048835.1 | - | 313 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB651299.1 | - | 3 | 313 | Chr. 17 - 69894579 - 69923995 [-] on Build GRCm38 | |

Gm36594
Genomic Map