siRNA Details
| Gene Symbol: Gm36594 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK048835.1 | - | 893 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB391307.2 | - | 1 | 98 | Chr. 17 - 69893765 - 69894415 [-] on Build GRCm38 | |

Gm36594
Genomic Map