siRNA Details
Gene Symbol: 9930111H07Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_108086.1 | 4 | 705 | |||
GenBank mRNA | AK020686.1 | - | 4 | 706 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 9930111H07Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_108086.1 | 4 | 705 | |||
GenBank mRNA | AK020686.1 | - | 4 | 706 |