siRNA Details
| Gene Symbol: 9930111H07Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_108086.1 | 2 | 79 | |||
| GenBank mRNA | AK020686.1 | - | 2 | 80 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY723014.1 | - | 2 | 80 | Chr. 1 - 85775271 - 85784505 [+] on Build GRCm38 | |

9930111H07Rik
Genomic Map