siRNA Details
| Gene Symbol: 9630015K15Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK020680.1 | - | 3, 4 | 369 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY722440.1 | - | 3, 4 | 369 | Chr. 14 - 64116314 - 64127599 [+] on Build GRCm38 | |

9630015K15Rik
Genomic Map