siRNA Details
| Gene Symbol: Gm12709 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040444.1 | 4 | 1397 | |||
| GenBank mRNA | AK038336.1 | - | 4 | 1398 | ||
| BC064118.1 | - | 4 | 1363 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm12709| Gene Symbol: Gm12709 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_040444.1 | 4 | 1397 | |||
| GenBank mRNA | AK038336.1 | - | 4 | 1398 | ||
| BC064118.1 | - | 4 | 1363 | |||