siRNA Details
| Gene Symbol: Gm34483 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK144767.1 | - | 473 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BY734350.1 | - | 4 | 473 | Chr. 2 - 102955347 - 102968714 [+] on Build GRCm38 | |

Gm34483
Genomic Map