siRNA Details
| Gene Symbol: Gm31324 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK133279.1 | - | 3 | 420 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB616655.1 | - | 3 | 420 | Chr. 2 - 48793390 - 48796510 [+] on Build GRCm38 | |

Gm31324
Genomic Map