siRNA Details
| Gene Symbol: Sgpp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK087798.1 | - | 1 | 70 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB666583.1 | - | 1 | 70 | Chr. 1 - 78327667 - 78340106 [-] on Build GRCm38 | |

Sgpp2
Genomic Map