siRNA Details
| Gene Symbol: Dreh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_105051.1 | 2 | 665 | |||
| GenBank mRNA | AK050349.1 | - | 2 | 666 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | AI451793.1 | - | 2 | 501 | Chr. 17 - 64767275 - 64767904 [+] on Build GRCm38 | |

Dreh
Genomic Map