siRNA Details
| Gene Symbol: Dreh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_105051.1 | 1 | 166 | |||
| GenBank mRNA | AK050349.1 | - | 1 | 167 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB653987.1 | - | 167 | Chr. - - [-] on Build | ||

Dreh
Genomic Map