siRNA Details
| Gene Symbol: 2310015A10Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033514.1 | 3 | 287 | |||
| GenBank mRNA | AK009348.1 | - | 3 | 276 | ||
| AK053779.1 | - | 3 | 257 | |||
| AK160272.1 | - | 3 | 288 | |||
| BB617891.1 | - | 3 | 288 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

2310015A10Rik| Gene Symbol: 2310015A10Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_033514.1 | 3 | 287 | |||
| GenBank mRNA | AK009348.1 | - | 3 | 276 | ||
| AK053779.1 | - | 3 | 257 | |||
| AK160272.1 | - | 3 | 288 | |||
| BB617891.1 | - | 3 | 288 | |||