siRNA Details
| Gene Symbol: 2310015A16Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK009349.1 | - | 2 | 339 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

2310015A16Rik| Gene Symbol: 2310015A16Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK009349.1 | - | 2 | 339 | ||