siRNA Details
| Gene Symbol: Zeb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK051204.1 | - | 1 | 1990 | ||
| AK051436.1 | - | 1 | 1989 | |||
| AK158356.1 | - | 1 | 343 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zeb2| Gene Symbol: Zeb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK051204.1 | - | 1 | 1990 | ||
| AK051436.1 | - | 1 | 1989 | |||
| AK158356.1 | - | 1 | 343 | |||