siRNA Details
Gene Symbol: 4930455H04Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040596.1 | 3 | 1079 | |||
GenBank mRNA | AK015465.1 | - | 3 | 1080 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 4930455H04Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040596.1 | 3 | 1079 | |||
GenBank mRNA | AK015465.1 | - | 3 | 1080 |