siRNA Details
| Gene Symbol: 4930455D15Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045381.1 | 5 | 1504 | |||
| GenBank mRNA | AK015461.1 | - | 5 | 1505 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4930455D15Rik| Gene Symbol: 4930455D15Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_045381.1 | 5 | 1504 | |||
| GenBank mRNA | AK015461.1 | - | 5 | 1505 | ||