siRNA Details
| Gene Symbol: 4931428F04Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK049007.1 | - | 3 | 580 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BB395172.2 | - | 3 | 615 | Chr. 8 - 105287914 - 105289504 [-] on Build GRCm38 | |

4931428F04Rik
Genomic Map