siRNA Details
| Gene Symbol: Gm5144 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC028532.1 | - | 2 | 564 | ||
| BC038676.1 | - | 2 | 577 | |||
| BC051480.1 | - | 2 | 585 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm5144| Gene Symbol: Gm5144 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC028532.1 | - | 2 | 564 | ||
| BC038676.1 | - | 2 | 577 | |||
| BC051480.1 | - | 2 | 585 | |||