siRNA Details
| Gene Symbol: C9orf62 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173520.2 | NP_775791.1 | 3 | 1546 | ||
| GenBank mRNA | BC034752.1 | - | 3 | 1546 | 5193354 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

C9orf62| Gene Symbol: C9orf62 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173520.2 | NP_775791.1 | 3 | 1546 | ||
| GenBank mRNA | BC034752.1 | - | 3 | 1546 | 5193354 | |