siRNA Details
Gene Symbol: C9orf139 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207511.2 | NP_997394.1 | 3 | 2027 | ||
XM_017014717.1 | XP_016870206.1 | 3 | 992 | |||
GenBank mRNA | AK093587.1 | - | 2 | 396 | ||
AK124899.1 | - | 3 | 2027 | |||
BC132809.1 | - | 2 | 801 | 40146831 | ||
BC136839.1 | - | 2 | 801 | 9020830 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000623196.1 | - | 2027 | Chr. - - [-] on Build |
