siRNA Details
| Gene Symbol: EML2-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_130728.1 | 3 | 580 | |||
| NR_130729.1 | 2 | 386 | ||||
| GenBank mRNA | BC000856.1 | - | 2 | 301 | ||
| BI334767.1 | - | 2 | 386 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

EML2-AS1| Gene Symbol: EML2-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_130728.1 | 3 | 580 | |||
| NR_130729.1 | 2 | 386 | ||||
| GenBank mRNA | BC000856.1 | - | 2 | 301 | ||
| BI334767.1 | - | 2 | 386 | |||