siRNA Details
| Gene Symbol: MIR202HG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_108078.1 | 3 | 349 | |||
| NR_108079.1 | 2 | 201 | ||||
| GenBank mRNA | HY017367.1 | - | 2 | 202 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

MIR202HG| Gene Symbol: MIR202HG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_108078.1 | 3 | 349 | |||
| NR_108079.1 | 2 | 201 | ||||
| GenBank mRNA | HY017367.1 | - | 2 | 202 | ||