siRNA Details
Gene Symbol: MIR202HG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_108078.1 | 2, 3 | 241 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | HY038901.1 | - | 2, 3 | 242 | Chr. 10 - 133246845 - 133247884 [-] on Build GRCh38 |
