siRNA Details
| Gene Symbol: PAX8-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015377.2 | 4, 5 | 975 | |||
| GenBank mRNA | AK001856.1 | - | 1, 2 | 123 | ||
| AK056355.1 | - | 4, 5 | 613 | |||
| AL390179.1 | - | 1, 2 | 133 | |||
| BC036699.2 | - | 4, 5 | 975 | |||
| BX537688.1 | - | 3, 4 | 516 | |||

PAX8-AS1
Genomic Map