siRNA Details
| Gene Symbol: PAX8-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_015377.2 | 5 | 1101 | |||
| GenBank mRNA | AK001856.1 | - | 2 | 249 | ||
| AK307781.1 | - | 4 | 449 | |||
| AL390179.1 | - | 2 | 259 | |||
| BC033562.1 | - | 4 | 984 | |||
| BC036699.2 | - | 5 | 1101 | |||
| BX537688.1 | - | 4 | 642 | |||

PAX8-AS1
Genomic Map