siRNA Details
Gene Symbol: LINC00449 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_133664.1 | 1 | 221 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000366259.2 | - | 1 | 210 | Chr. 7 - 72893318 - 72893594 [-] on Build GRCh38 |
