siRNA Details
| Gene Symbol: LOC105372582 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135006.1 | 1 | 105 | |||
| GenBank mRNA | DB215985.1 | - | 1 | 105 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC105372582| Gene Symbol: LOC105372582 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135006.1 | 1 | 105 | |||
| GenBank mRNA | DB215985.1 | - | 1 | 105 | ||