siRNA Details
| Gene Symbol: ITPR1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_108075.1 | 2 | 783 | |||
| GenBank mRNA | AL832729.1 | - | 1 | 2208 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ITPR1-AS1| Gene Symbol: ITPR1-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_108075.1 | 2 | 783 | |||
| GenBank mRNA | AL832729.1 | - | 1 | 2208 | ||