siRNA Details
| Gene Symbol: LOC101929574 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120661.1 | 1 | 205 | |||
| GenBank mRNA | BI439066.1 | - | 1 | 205 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | CK818365.1 | - | 1 | 195 | Chr. 10 - 80529632 - 80535932 [-] on Build GRCh38 | |

LOC101929574
Genomic Map