siRNA Details
| Gene Symbol: CSE1L-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110624.1 | 2 | 221 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000417781.5 | - | 2 | 209 | Chr. 20 - 49040463 - 49046044 [-] on Build GRCh38 | |

CSE1L-AS1
Genomic Map