siRNA Details
Gene Symbol: DPP10-AS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_132105.1 | 2 | 571 | |||
GenBank mRNA | BC033566.1 | - | 2 | 571 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000417844.1 | - | 2 | 571 | Chr. 2 - 114828432 - 114833548 [-] on Build GRCh38 |
