siRNA Details
| Gene Symbol: LOC101927365 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_125843.1 | 3 | 730 | |||
| GenBank mRNA | BI913790.1 | - | 2 | 478 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101927365| Gene Symbol: LOC101927365 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_125843.1 | 3 | 730 | |||
| GenBank mRNA | BI913790.1 | - | 2 | 478 | ||