siRNA Details
Gene Symbol: LOC101927365 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_125843.1 | 3 | 638 | |||
GenBank mRNA | BI913790.1 | - | 2 | 386 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000418945.1 | - | 2 | 1207 | Chr. 6 - 99424922 - 99431373 [+] on Build GRCh38 |
