siRNA Details
| Gene Symbol: MKX-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_121652.1 | 2 | 378 | |||
| GenBank mRNA | BG194628.1 | - | 1 | 50 | ||
| BU195543.1 | - | 2 | 215 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

MKX-AS1| Gene Symbol: MKX-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_121652.1 | 2 | 378 | |||
| GenBank mRNA | BG194628.1 | - | 1 | 50 | ||
| BU195543.1 | - | 2 | 215 | |||