siRNA Details
Gene Symbol: TM4SF19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | CA430273.1 | - | 1 | 50 |
Gene Symbol: TM4SF19-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_121665.1 | 2 | 224 | |||
GenBank mRNA | CK819994.1 | - | 2 | 224 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000420226.1 | - | 2 | 226 | Chr. 3 - 196318330 - 196323673 [+] on Build GRCh38 |
