siRNA Details
| Gene Symbol: PDCD4-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_026932.1 | 1 | 763 | |||
| GenBank mRNA | AK093606.1 | - | 1 | 763 | ||
| AK097703.1 | - | 1 | 1638 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000420367.1 | - | 2 | 504 | Chr. 10 - 110869868 - 110872233 [-] on Build GRCh38 | |

PDCD4-AS1
Genomic Map