siRNA Details
| Gene Symbol: LINC01510 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120506.1 | 5 | 883 | |||
| GenBank mRNA | BG189453.1 | - | 2 | 218 | ||
| BG195638.1 | - | 2 | 217 | |||
| BG213277.1 | - | 2 | 220 | |||
| LN812953.1 | - | 4 | 544 | |||
| LN812954.1 | - | 5 | 848 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GenBankmRNA | BG184957.1 | - | 2 | 216 | Chr. 7 - 116563594 - 116571229 [-] on Build GRCh38 | |

LINC01510
Genomic Map